accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClPClOO

CAS RN 770-12-7

IUPAC Name: phenyl chlorophosphonochloridate
Synonyms: phenyl dichlorophosphate
Molecular Weight (g/mol): 210.98
Molecular Formula: C6H5Cl2O2P
InChi Key: TXFOLHZMICYNRM-UHFFFAOYSA-N
SMILES: ClP(Cl)(=O)OC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (3)
  • (3)
  • (1)
  • (1)
Boiling Point 
  • (5)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (5)
  • (3)
  • (3)
  • (1)
  • (1)
  • (5)
  • (3)
  • (2)
  • (2)