accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 770-30-9

H3CSNNH2NN

CAS RN 770-30-9

IUPAC Name: 4-amino-2-(methylsulfanyl)pyrimidine-5-carbonitrile
Synonyms: 4-amino-2-(methylsulfanyl)pyrimidine-5-carbonitrile
Molecular Weight (g/mol): 166.2
Molecular Formula: C6H6N4S
InChi Key: VYDHBDPZNLVELI-UHFFFAOYSA-N
SMILES: CSC1=NC=C(C#N)C(N)=N1
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)