accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COHO

CAS RN 770-35-4

IUPAC Name: 1-phenoxypropan-2-ol
Synonyms: 1-phenoxy-2-propanol 2PROPANOL1phenoxy phenoxyisopropanol 1-phenoxypropan-2-ol (8)
Molecular Weight (g/mol): 152.19
Molecular Formula: C9H12O2
InChi Key: IBLKWZIFZMJLFL-UHFFFAOYNA-N
SMILES: CC(O)COC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results