CAS RN 7704-34-9
Sulfur, 99.5+%, refined
CAS: 7704-34-9 Molecular Formula: S Molecular Weight (g/mol): 32.06 MDL Number: MFCD00085316 InChI Key: NINIDFKCEFEMDL-UHFFFAOYSA-N Synonym: polymeric,lac,ad usum externum,acmc-20a47y,rubbervulcanizationagentis ot PubChem CID: 5362487 ChEBI: CHEBI:26833 IUPAC Name: sulfur SMILES: [S]
Sulfur, 99.5%, sublimed
CAS: 7704-34-9 Molecular Formula: S Molecular Weight (g/mol): 32.06 MDL Number: MFCD00085316 InChI Key: NINIDFKCEFEMDL-UHFFFAOYSA-N Synonym: polymeric,lac,ad usum externum,acmc-20a47y,rubbervulcanizationagentis ot PubChem CID: 5362487 ChEBI: CHEBI:26833 IUPAC Name: sulfur SMILES: [S]
Sulfur powder, sublimed, -100 mesh, 99.5%
CAS: 7704-34-9 Molecular Formula: S Molecular Weight (g/mol): 32.06 MDL Number: MFCD00085316 InChI Key: NINIDFKCEFEMDL-UHFFFAOYSA-N Synonym: polymeric,lac,ad usum externum,acmc-20a47y,rubbervulcanizationagentis ot PubChem CID: 5362487 ChEBI: CHEBI:26833 IUPAC Name: sulfur SMILES: [S]
Sulfur, flowers, 99%
CAS: 7704-34-9 Molecular Formula: S
Sulfur powder, -325 mesh, 99.5%
CAS: 7704-34-9 Molecular Formula: S Molecular Weight (g/mol): 32.06 MDL Number: MFCD00085316 InChI Key: NINIDFKCEFEMDL-UHFFFAOYSA-N Synonym: polymeric,lac,ad usum externum,acmc-20a47y,rubbervulcanizationagentis ot PubChem CID: 5362487 ChEBI: CHEBI:26833 IUPAC Name: sulfur SMILES: [S]
Sulfur powder, precipitated, 99.5%
CAS: 7704-34-9 Molecular Formula: S Molecular Weight (g/mol): 32.06 MDL Number: MFCD00085316 InChI Key: NINIDFKCEFEMDL-UHFFFAOYSA-N Synonym: polymeric,lac,ad usum externum,acmc-20a47y,rubbervulcanizationagentis ot PubChem CID: 5362487 ChEBI: CHEBI:26833 IUPAC Name: sulfur SMILES: [S]
Sulfur pieces, Puratronic™, 99.9995% (metals basis)
CAS: 7704-34-9 Molecular Formula: S Molecular Weight (g/mol): 32.06 MDL Number: MFCD00085316 InChI Key: NINIDFKCEFEMDL-UHFFFAOYSA-N Synonym: polymeric,lac,ad usum externum,acmc-20a47y,rubbervulcanizationagentis ot PubChem CID: 5362487 ChEBI: CHEBI:26833 IUPAC Name: sulfur SMILES: [S]
Sulphur solution for ICP/MS 10mg/L H2O matrix, Fisher chemical™
CAS: 7704-34-9 Molecular Formula: S Molecular Weight (g/mol): 32.06 MDL Number: MFCD00085316 InChI Key: NINIDFKCEFEMDL-UHFFFAOYSA-N PubChem CID: 5362487 ChEBI: CHEBI:26833 IUPAC Name: sulfur SMILES: [S]
Sulfur pieces, Puratronic™, 99.999% (metals basis)
CAS: 7704-34-9 Molecular Formula: S Molecular Weight (g/mol): 32.06 MDL Number: MFCD00085316 InChI Key: NINIDFKCEFEMDL-UHFFFAOYSA-N Synonym: polymeric,lac,ad usum externum,acmc-20a47y,rubbervulcanizationagentis ot PubChem CID: 5362487 ChEBI: CHEBI:26833 IUPAC Name: sulfur SMILES: [S]
Sulfur Standard: S @ 10 μg/g in B100 Biodiesel, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Sulfur - S @ 3,500ug/g in Residual Oil, 500mL
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Sulfur Standard: S @ 30000 μg/g (3.00 wt%) in Residual Oil
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications