accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7704-98-5

TiHH

CAS RN 7704-98-5

IUPAC Name: titanium(2+) dihydride
Synonyms: titanium hydride
Molecular Weight (g/mol): 49.88
Molecular Formula: H2Ti
InChi Key: KAZWGWWZKAHTKC-UHFFFAOYSA-N
SMILES: [H-].[H-].[Ti++]
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (7)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (7)
  • (2)
  • (1)