CAS RN 7704-99-6
IUPAC Name:
λ²-zirconium(2+) dihydride
Synonyms:
λ²-zirconium(2+) dihydride
Molecular Weight (g/mol):
93.24
Molecular Formula:
H2Zr
InChi Key:
IUJYJRCMJZIAJY-UHFFFAOYSA-N
SMILES:
[H-].[H-].[Zr++]
1
–
3
of
3
results
1
Zirconium hydride, 99%, Thermo Scientific Chemicals
CAS: 7704-99-6 MDL Number: MFCD00148878