accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 771-15-3

IUPAC Name: 2-bromonaphthalen-1-ol
Synonyms: 2-bromonaphthalen-1-ol
Molecular Weight (g/mol): 223.07
Molecular Formula: C10H7BrO
InChi Key: LOHOSHDZQVKDPS-UHFFFAOYSA-N
SMILES: OC1=C2C=CC=CC2=CC=C1Br
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results