accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 771-61-9

IUPAC Name: pentafluorophenol
Synonyms: pentafluorophenol
Molecular Weight (g/mol): 184.07
Molecular Formula: C6HF5O
InChi Key: XBNGYFFABRKICK-UHFFFAOYSA-N
SMILES: OC1=C(F)C(F)=C(F)C(F)=C1F
Molecular Weight (g/mol) 
  • (5)
  • (9)
Quantity 
  • (2)
  • (3)
  • (2)
  • (4)
Percent Purity 
  • (4)
  • (3)
  • (3)
Concentration 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (3)
  • (3)
  • (1)
  • (4)
  • (5)
  • (9)
  • (2)
  • (3)
  • (2)
  • (4)
  • (4)
  • (3)
  • (3)
  • (6)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)