accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2NH2N

CAS RN 771-97-1

IUPAC Name: naphthalene-2,3-diamine
Synonyms: 2,3-naphthalenediamine
Molecular Weight (g/mol): 158.2
Molecular Formula: C10H10N2
InChi Key: XTBLDMQMUSHDEN-UHFFFAOYSA-N
SMILES: NC1=C(N)C=C2C=CC=CC2=C1
Molecular Weight (g/mol) 
  • (3)
  • (6)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (3)
  • (6)
  • (3)
  • (6)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)