CAS RN 77128-72-4
IUPAC Name:
2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(4-methoxyphenyl)propanoic acid
Synonyms:
2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(4-methoxyphenyl)propanoic acid
Molecular Weight (g/mol):
417.46
Molecular Formula:
C25H23NO5
InChi Key:
JYQODLWFOPCSCS-UHFFFAOYNA-N
SMILES:
COC1=CC=C(CC(NC(=O)OCC2C3=CC=CC=C3C3=CC=CC=C23)C(O)=O)C=C1
1
–
1
of
1
results
1
N-Fmoc-4-methoxy-L-phenylalanine, 95%
CAS: 77128-72-4 Molecular Formula: C25H23NO5 Molecular Weight (g/mol): 417.461 MDL Number: MFCD00153368 InChI Key: JYQODLWFOPCSCS-QHCPKHFHSA-N Synonym: fmoc-tyr me-oh,fmoc-o-methyl-l-tyrosine,fmoc-4-methoxy-l-phenylalanine,fmoc-l-4-methoxyphenylalanine,fmoc-4-methoxy-phe-oh,fmoc-o-methyl-tyr-oh,fmoc-l-tyr me,fmoc-4-methoxy-l-phe-oh,fmoc-l-4-methoxyphe PubChem CID: 2762281 IUPAC Name: (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-methoxyphenyl)propanoic acid SMILES: COC1=CC=C(C=C1)CC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24