accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 77145-39-2

NNH2NHCl

CAS RN 77145-39-2

IUPAC Name: 1-(pyridin-2-yl)piperidin-4-amine hydrochloride
Synonyms: 1-(pyridin-2-yl)piperidin-4-amine hydrochloride
Molecular Weight (g/mol): 213.71
Molecular Formula: C10H16ClN3
InChi Key: HNICPQFPDAEZKW-UHFFFAOYSA-N
SMILES: Cl.NC1CCN(CC1)C1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results