accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COONNCl

CAS RN 77173-67-2

IUPAC Name: ethyl 4-chloro-8-cyanoquinoline-3-carboxylate
Synonyms: ethyl 4-chloro-8-cyanoquinoline-3-carboxylate
Molecular Weight (g/mol): 260.68
Molecular Formula: C13H9ClN2O2
InChi Key: MHOFAUCLSGVKOH-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN=C2C(=CC=CC2=C1Cl)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results