CAS RN 77182-82-2
CAS RN 77182-82-2
IUPAC Name:
ammonium 2-amino-4-[hydroxy(methyl)phosphoryl]butanoate
Synonyms:
glufosinate-ammonium glufosinate ammonium (iso)
Molecular Weight (g/mol):
198.16
Molecular Formula:
C5H15N2O4P
InChi Key:
ZBMRKNMTMPPMMK-UHFFFAOYNA-N
SMILES:
[NH4+].CP(O)(=O)CCC(N)C([O-])=O
1
–
5
of
5
results
1
Glufosinate ammonium, 95%, Thermo Scientific™
CAS: 77182-82-2 Molecular Formula: C5H15N2O4P Molecular Weight (g/mol): 198.16 MDL Number: MFCD00055562,MFCD00211362 InChI Key: ZBMRKNMTMPPMMK-UHFFFAOYNA-N Synonym: DL-Phosphinothricin; 2-Amino-4-(hydroxymethylphosphinyl)butyric acid ammonium salt PubChem CID: 122130249 IUPAC Name: ammonium 2-amino-4-[hydroxy(methyl)phosphoryl]butanoate SMILES: [NH4+].CP(O)(=O)CCC(N)C([O-])=O
3
Glufosinate ammonium 100 μg/mL in Acetonitrile:Water, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality