CAS RN 7722-64-7
CAS RN 7722-64-7
IUPAC Name:
potassium manganesoylolate
Synonyms:
potassium permanganate 高锰酸钾
Molecular Weight (g/mol):
158.03
Molecular Formula:
KMnO4
InChi Key:
VZJVWSHVAAUDKD-UHFFFAOYSA-N
SMILES:
[K+].[O-][Mn](=O)(=O)=O
1
–
3
of
3
results
2
Potassium Permanganate, Analytical Grade, Penta Chemicals
CAS: 7722-64-7 Molecular Weight (g/mol): 158.04
3
Potassium Permanganate, Pure Grade, Penta Chemicals
CAS: 7722-64-7 Molecular Weight (g/mol): 158.04