accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH4OHPOHOO

CAS RN 7722-76-1

IUPAC Name: ammonium dihydrogen phosphate
Synonyms: monoammonium phosphate ammonium biphosphate monoammonium dihydrogen orthophosphate primary ammonium phosphate phosphoric acid, monoammonium salt monoammonium acid phosphate ammonium monophosphate ammonium orthophosphate dihydrogen ammonium dihydrogen orthophosphate monoammonium hydrogen phosphate
Molecular Weight (g/mol): 115.03
Molecular Formula: H6NO4P
InChi Key: LFVGISIMTYGQHF-UHFFFAOYSA-N
SMILES: [NH4+].OP(O)([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (50)
Quantity 
  • (7)
  • (1)
  • (1)
  • (6)
  • (1)
  • (1)
  • (5)
  • (3)
  • (8)
  • (1)
  • (1)
  • (12)
Percent Purity 
  • (8)
  • (1)
  • (4)
  • (5)
  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (3)
  • (2)
  • (3)
Grade 
  • (2)
  • (5)
  • (3)
  • (5)
  • (6)
  • (1)
  • (2)
  • (2)
  • (2)
  • (6)
  • (2)
Physical Form 
  • (4)
  • (3)
  • (1)
  • (6)
  • (7)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (6)
  • (9)
  • (1)
  • (6)
  • (1)
  • (14)
  • (7)
  • (3)
  • (5)
  • (1)
  • (3)
  • (50)
  • (7)
  • (1)
  • (1)
  • (6)
  • (1)
  • (1)
  • (5)
  • (3)
  • (8)
  • (1)
  • (1)
  • (12)
  • (8)
  • (1)
  • (4)
  • (5)
  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (5)
  • (3)
  • (5)
  • (6)
  • (1)
  • (2)
  • (2)
  • (2)
  • (6)
  • (2)
  • (4)
  • (3)
  • (1)
  • (6)
  • (7)
  • (4)
  • (6)
  • (6)