accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrBr

CAS RN 7726-95-6

IUPAC Name: bromide
Synonyms: bromide caswell no. 112 inorganic bromides bromine anion caswell no. 499D bromide ions bromine liquid BrR RBr br- element
Molecular Weight (g/mol): 79.91
Molecular Formula: Br
InChi Key: CPELXLSAUQHCOX-UHFFFAOYSA-M
SMILES: [Br-]
Quantity 
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)
  • (7)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (4)
  • (2)
  • (2)
  • (4)
  • (3)
  • (4)
  • (2)
Grade 
  • (4)
  • (4)
  • (3)
  • (3)
  • (3)
  • (4)
Physical Form 
  • (1)
  • (3)
  • (14)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (4)
  • (1)
  • (4)
  • (3)
  • (1)
  • (12)
  • (10)
  • (4)
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)
  • (7)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)
  • (1)
  • (2)
  • (1)
  • (4)
  • (2)
  • (2)
  • (4)
  • (3)
  • (4)
  • (2)
  • (4)
  • (4)
  • (3)
  • (3)
  • (3)
  • (4)
  • (1)
  • (3)
  • (14)
  • (4)
  • (4)
  • (4)