accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH4NH4OSOOOOSOOO

CAS RN 7727-54-0

IUPAC Name: diammonium [(sulfonatoperoxy)sulfonyl]oxidanide
Synonyms: ammonium persulfate
Molecular Weight (g/mol): 228.19
Molecular Formula: H8N2O8S2
InChi Key: ROOXNKNUYICQNP-UHFFFAOYSA-N
SMILES: [NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O
Molecular Weight (g/mol) 
  • (46)
  • (5)
Quantity 
  • (6)
  • (3)
  • (12)
  • (3)
  • (1)
  • (2)
  • (7)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (7)
  • (1)
  • (1)
Physical Form 
  • (8)
  • (9)
  • (2)
  • (3)
  • (4)
  • (7)
  • (7)
Grade 
  • (6)
  • (11)
  • (3)
  • (1)
  • (6)
  • (3)
  • (5)
  • (1)
  • (3)
Percent Purity 
  • (17)
  • (7)
  • (4)
  • (9)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (3)
  • (8)
  • (1)
  • (5)
  • (2)
  • (3)
  • (11)
  • (8)
  • (3)
  • (46)
  • (5)
  • (6)
  • (3)
  • (12)
  • (3)
  • (1)
  • (2)
  • (7)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (7)
  • (1)
  • (1)
  • (8)
  • (9)
  • (2)
  • (3)
  • (4)
  • (7)
  • (7)
  • (6)
  • (11)
  • (3)
  • (1)
  • (6)
  • (3)
  • (5)
  • (1)
  • (3)
  • (17)
  • (7)
  • (4)
  • (9)