accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFFNFF

CAS RN 773-82-0

IUPAC Name: pentafluorobenzonitrile
Synonyms: benzonitrile, pentafluoro-
Molecular Weight (g/mol): 193.08
Molecular Formula: C7F5N
InChi Key: YXWJGZQOGXGSSC-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(C#N)C(F)=C1F
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)