accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHHOI

CAS RN 773869-57-1

IUPAC Name: 5-(hydroxymethyl)-2-iodophenol
Synonyms: 5-(hydroxymethyl)-2-iodophenol
Molecular Weight (g/mol): 250.04
Molecular Formula: C7H7IO2
InChi Key: VIWWSAGABDIIFF-UHFFFAOYSA-N
SMILES: OCC1=CC(O)=C(I)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results