CAS RN 773872-97-2
CAS RN 773872-97-2
IUPAC Name:
[2-(thiophen-2-yl)phenyl]methanol
Synonyms:
[2-(thiophen-2-yl)phenyl]methanol
Molecular Weight (g/mol):
190.26
Molecular Formula:
C11H10OS
InChi Key:
IUULMJAEIKVTKZ-UHFFFAOYSA-N
SMILES:
OCC1=CC=CC=C1C1=CC=CS1
1
–
1
of
1
results
1
(2-Thien-2-ylphenyl)methanol, 97%, Thermo Scientific™
CAS: 773872-97-2 Molecular Formula: C11H10OS Molecular Weight (g/mol): 190.26 MDL Number: MFCD06203081 InChI Key: IUULMJAEIKVTKZ-UHFFFAOYSA-N Synonym: 2-thien-2-ylphenyl methanol,2-thiophen-2-yl phenyl methanol,2-thien-2-yl phenyl methanol,2-thiophen-2-ylphenyl methanol,2-2-thienyl phenyl methanol,benzenemethanol,2-2-thienyl,2-2-thienyl phenyl methan-1-ol PubChem CID: 2795559 SMILES: OCC1=CC=CC=C1C1=CC=CS1