accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2Si

CAS RN 775-12-2

IUPAC Name: diphenylsilane
Synonyms: diphenylsilane
Molecular Weight (g/mol): 184.31
Molecular Formula: C12H12Si
InChi Key: VDCSGNNYCFPWFK-UHFFFAOYSA-N
SMILES: [SiH2](C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)