accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7752-82-1

NH2NBrN

CAS RN 7752-82-1

IUPAC Name: 5-bromopyrimidin-2-amine
Synonyms: 5-bromopyrimidin-2-amine
Molecular Weight (g/mol): 174
Molecular Formula: C4H4BrN3
InChi Key: UHRHPPKWXSNZLR-UHFFFAOYSA-N
SMILES: NC1=NC=C(Br)C=N1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (3)
  • (1)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (5)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)