CAS RN 7757-79-1
Potassium nitrate, 99+%, for analysis
CAS: 7757-79-1 Molecular Formula: KNO3 Molecular Weight (g/mol): 101.10 MDL Number: MFCD00011409 InChI Key: FGIUAXJPYTZDNR-UHFFFAOYSA-N Synonym: potassium nitrate,saltpeter,nitre,nitrate of potash,vicknite,nitric acid potassium salt,nitric acid, potassium salt,saltpetre,kaliumnitrat,kalii nitras PubChem CID: 24434 ChEBI: CHEBI:63043 SMILES: [K+].[O-][N+]([O-])=O
Potassium nitrate, 99%, extra pure
CAS: 7757-79-1 Molecular Formula: KNO3 Molecular Weight (g/mol): 101.10 MDL Number: MFCD00011409 InChI Key: FGIUAXJPYTZDNR-UHFFFAOYSA-N Synonym: potassium nitrate,saltpeter,nitre,nitrate of potash,vicknite,nitric acid potassium salt,nitric acid, potassium salt,saltpetre,kaliumnitrat,kalii nitras PubChem CID: 24434 ChEBI: CHEBI:63043 SMILES: [K+].[O-][N+]([O-])=O
Potassium Nitrate, Technical, Fisher Chemical™
CAS: 7757-79-1 Molecular Formula: KNO3 Molecular Weight (g/mol): 101.10 MDL Number: MFCD00011409 InChI Key: FGIUAXJPYTZDNR-UHFFFAOYSA-N Synonym: potassium nitrate,saltpeter,nitre,nitrate of potash,vicknite,nitric acid potassium salt,nitric acid, potassium salt,saltpetre,kaliumnitrat,kalii nitras PubChem CID: 24434 ChEBI: CHEBI:63043 SMILES: [K+].[O-][N+]([O-])=O
Potassium nitrate, 99+%, ACS reagent, Thermo Scientific™
CAS: 7757-79-1 Molecular Formula: KNO3 Molecular Weight (g/mol): 101.1 MDL Number: MFCD00011409 InChI Key: FGIUAXJPYTZDNR-UHFFFAOYSA-N Synonym: potassium nitrate,saltpeter,nitre,nitrate of potash,vicknite,nitric acid potassium salt,nitric acid, potassium salt,saltpetre,kaliumnitrat,kalii nitras PubChem CID: 24434 ChEBI: CHEBI:63043 IUPAC Name: potassium;nitrate SMILES: [N+](=O)([O-])[O-].[K+]
Potassium nitrate, 99%, Thermo Scientific™
CAS: 7757-79-1 Molecular Formula: KNO3 Molecular Weight (g/mol): 101.102 MDL Number: MFCD00011409 InChI Key: FGIUAXJPYTZDNR-UHFFFAOYSA-N Synonym: potassium nitrate,saltpeter,nitre,nitrate of potash,vicknite,nitric acid potassium salt,nitric acid, potassium salt,saltpetre,kaliumnitrat,kalii nitras PubChem CID: 24434 ChEBI: CHEBI:63043 IUPAC Name: potassium;nitrate SMILES: [N+](=O)([O-])[O-].[K+]
Potassium, plasma standard solution, Specpure™ K 1000μg/mL
CAS: 7757-79-1 Molecular Formula: KNO3 Molecular Weight (g/mol): 101.10 MDL Number: MFCD00011409 InChI Key: FGIUAXJPYTZDNR-UHFFFAOYSA-N PubChem CID: 24434 ChEBI: CHEBI:63043 SMILES: [K+].[O-][N+]([O-])=O
Potassium, plasma standard solution, Specpure™ K 10,000μg/mL
CAS: 7757-79-1 Molecular Formula: KNO3 Molecular Weight (g/mol): 101.10 MDL Number: MFCD00011409 InChI Key: FGIUAXJPYTZDNR-UHFFFAOYSA-N PubChem CID: 24434 ChEBI: CHEBI:63043 SMILES: [K+].[O-][N+]([O-])=O
Potassium nitrate, 99.994% (metals basis), Thermo Scientific™
CAS: 7757-79-1 Molecular Formula: KNO3 Molecular Weight (g/mol): 101.102 MDL Number: MFCD00011409 InChI Key: FGIUAXJPYTZDNR-UHFFFAOYSA-N Synonym: potassium nitrate,saltpeter,nitre,nitrate of potash,vicknite,nitric acid potassium salt,nitric acid, potassium salt,saltpetre,kaliumnitrat,kalii nitras PubChem CID: 24434 ChEBI: CHEBI:63043 IUPAC Name: potassium;nitrate SMILES: [N+](=O)([O-])[O-].[K+]
Potassium nitrate, ACS, 99.0% min, Thermo Scientific™
CAS: 7757-79-1 Molecular Formula: KNO3 Molecular Weight (g/mol): 101.102 MDL Number: MFCD00011409 InChI Key: FGIUAXJPYTZDNR-UHFFFAOYSA-N Synonym: potassium nitrate,saltpeter,nitre,nitrate of potash,vicknite,nitric acid potassium salt,nitric acid, potassium salt,saltpetre,kaliumnitrat,kalii nitras PubChem CID: 24434 ChEBI: CHEBI:63043 IUPAC Name: potassium;nitrate SMILES: [N+](=O)([O-])[O-].[K+]
Potassium nitrate, Puratronic™, 99.995% (metals basis), Thermo Scientific™
CAS: 7757-79-1 Molecular Formula: IK Molecular Weight (g/mol): 166.003 MDL Number: MFCD00011409 InChI Key: NLKNQRATVPKPDG-UHFFFAOYSA-M Synonym: potassium iodide,potassium iodide ki,pima,kali iodide,thyro-block,knollide,kisol,asmofug e,potassium diiodide,potassium monoiodide PubChem CID: 4875 ChEBI: CHEBI:8346 IUPAC Name: potassium;iodide SMILES: [K+].[I-]
Potassium Nitrate, Extra Pure, SLR, Fisher Chemical™
CAS: 7757-79-1 Molecular Formula: KNO3 Molecular Weight (g/mol): 101.10 MDL Number: MFCD00011409 InChI Key: FGIUAXJPYTZDNR-UHFFFAOYSA-N Synonym: potassium nitrate,saltpeter,nitre,nitrate of potash,vicknite,nitric acid potassium salt,nitric acid, potassium salt,saltpetre,kaliumnitrat,kalii nitras PubChem CID: 24434 ChEBI: CHEBI:63043 SMILES: [K+].[O-][N+]([O-])=O
Potassium nitrate, 99.999%, (trace metal basis), Thermo Scientific™
CAS: 7757-79-1 Molecular Formula: KNO3 Molecular Weight (g/mol): 101.1 MDL Number: MFCD00011409 InChI Key: FGIUAXJPYTZDNR-UHFFFAOYSA-N Synonym: potassium nitrate,saltpeter,nitre,nitrate of potash,vicknite,nitric acid potassium salt,nitric acid, potassium salt,saltpetre,kaliumnitrat,kalii nitras PubChem CID: 24434 ChEBI: CHEBI:63043 IUPAC Name: potassium;nitrate SMILES: [N+](=O)([O-])[O-].[K+]
Potassium Nitrate, (Crystalline/ACS Reagent Grade), MP Biomedicals
CAS: 7757-79-1 Molecular Formula: KNO3 Molecular Weight (g/mol): 101.10 MDL Number: MFCD00011409 InChI Key: FGIUAXJPYTZDNR-UHFFFAOYSA-N Synonym: potassium nitrate,saltpeter,nitre,nitrate of potash,vicknite,nitric acid potassium salt,nitric acid, potassium salt,saltpetre,kaliumnitrat,kalii nitras PubChem CID: 24434 ChEBI: CHEBI:63043 IUPAC Name: potassium nitrate SMILES: [K+].[O-][N+]([O-])=O
Potassium nitrate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99%, Solstice
CAS: 7757-79-1 Molecular Formula: KNO3 Molecular Weight (g/mol): 101.10 MDL Number: MFCD00011409 InChI Key: FGIUAXJPYTZDNR-UHFFFAOYSA-N Synonym: potassium nitrate,saltpeter,nitre,nitrate of potash,vicknite,nitric acid potassium salt,nitric acid, potassium salt,saltpetre,kaliumnitrat,kalii nitras PubChem CID: 24434 ChEBI: CHEBI:63043 SMILES: [K+].[O-][N+]([O-])=O
- 1
- 2