CAS RN 7757-83-7
Sodium Sulfite Anhydrous, Certified AR for Analysis, Fisher Chemical™
CAS: 7757-83-7 Molecular Formula: Na2O3S Molecular Weight (g/mol): 126.037 MDL Number: 3503 InChI Key: GEHJYWRUCIMESM-UHFFFAOYSA-L Synonym: sodium sulfite,disodium sulfite,sodium sulphite,sodium sulfite anhydrous,sulfurous acid, disodium salt,sulftech,natrium sulfurosum,sodiumsulfite,natriumsulfid,natriumsulfit PubChem CID: 24437 ChEBI: CHEBI:86477 IUPAC Name: disodium;sulfite SMILES: [O-]S(=O)[O-].[Na+].[Na+]
Sodium sulfite, 98.5%, for analysis, anhydrous
CAS: 7757-83-7 Molecular Formula: Na2O3S Molecular Weight (g/mol): 126.04 MDL Number: MFCD00003503 InChI Key: GEHJYWRUCIMESM-UHFFFAOYSA-L Synonym: sodium sulfite,disodium sulfite,sodium sulphite,sodium sulfite anhydrous,sulfurous acid, disodium salt,sulftech,natrium sulfurosum,sodiumsulfite,natriumsulfid,natriumsulfit PubChem CID: 24437 ChEBI: CHEBI:86477 IUPAC Name: disodium;sulfite SMILES: [O-]S(=O)[O-].[Na+].[Na+]
Sodium sulfite, 97%, extra pure, anhydrous
CAS: 7757-83-7 Molecular Formula: Na2O3S Molecular Weight (g/mol): 126.04 InChI Key: GEHJYWRUCIMESM-UHFFFAOYSA-L Synonym: sodium sulfite,disodium sulfite,sodium sulphite,sodium sulfite anhydrous,sulfurous acid, disodium salt,sulftech,natrium sulfurosum,sodiumsulfite,natriumsulfid,natriumsulfit PubChem CID: 24437 ChEBI: CHEBI:86477 IUPAC Name: disodium;sulfite SMILES: [O-]S(=O)[O-].[Na+].[Na+]
Sodium sulfite, 98+%, ACS reagent, anhydrous
CAS: 7757-83-7 Molecular Formula: Na2O3S Molecular Weight (g/mol): 126.04 MDL Number: MFCD00003503 InChI Key: GEHJYWRUCIMESM-UHFFFAOYSA-L Synonym: sodium sulfite,disodium sulfite,sodium sulphite,sodium sulfite anhydrous,sulfurous acid, disodium salt,sulftech,natrium sulfurosum,sodiumsulfite,natriumsulfid,natriumsulfit PubChem CID: 24437 ChEBI: CHEBI:86477 IUPAC Name: disodium;sulfite SMILES: [O-]S(=O)[O-].[Na+].[Na+]
Sodium sulfite, 98%, for biochemistry
CAS: 7757-83-7 Molecular Formula: Na2O3S Molecular Weight (g/mol): 126.04 InChI Key: GEHJYWRUCIMESM-UHFFFAOYSA-L Synonym: sodium sulfite,disodium sulfite,sodium sulphite,sodium sulfite anhydrous,sulfurous acid, disodium salt,sulftech,natrium sulfurosum,sodiumsulfite,natriumsulfid,natriumsulfit PubChem CID: 24437 ChEBI: CHEBI:86477 IUPAC Name: disodium;sulfite SMILES: [O-]S(=O)[O-].[Na+].[Na+]
Sodium sulfite, anhydrous, 98%
CAS: 7757-83-7 Molecular Formula: Na2O3S Molecular Weight (g/mol): 126.037 MDL Number: MFCD00003503 InChI Key: GEHJYWRUCIMESM-UHFFFAOYSA-L Synonym: sodium sulfite,disodium sulfite,sodium sulphite,sodium sulfite anhydrous,sulfurous acid, disodium salt,sulftech,natrium sulfurosum,sodiumsulfite,natriumsulfid,natriumsulfit PubChem CID: 24437 ChEBI: CHEBI:86477 IUPAC Name: disodium;sulfite SMILES: [O-]S(=O)[O-].[Na+].[Na+]
Sodium sulfite, ACS, 98.0% min
CAS: 7757-83-7 | Na{2}SO{3}
Sodium sulfite, ACS, 98.0% min
CAS: 7757-83-7 Molecular Formula: Na2O3S Molecular Weight (g/mol): 126.037 MDL Number: MFCD00003503 InChI Key: GEHJYWRUCIMESM-UHFFFAOYSA-L Synonym: sodium sulfite,disodium sulfite,sodium sulphite,sodium sulfite anhydrous,sulfurous acid, disodium salt,sulftech,natrium sulfurosum,sodiumsulfite,natriumsulfid,natriumsulfit PubChem CID: 24437 ChEBI: CHEBI:86477 IUPAC Name: disodium;sulfite SMILES: [O-]S(=O)[O-].[Na+].[Na+]
Sodium Sulfite, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Sodium Sulfite, ≥98.0%, ACS Reagent, Solstice
CAS: 7757-83-7 Molecular Formula: Na2O3S Molecular Weight (g/mol): 126.037 MDL Number: MFCD00003503 InChI Key: GEHJYWRUCIMESM-UHFFFAOYSA-L Synonym: sodium sulfite,disodium sulfite,sodium sulphite,sodium sulfite anhydrous,sulfurous acid, disodium salt,sulftech,natrium sulfurosum,sodiumsulfite,natriumsulfid,natriumsulfit PubChem CID: 24437 ChEBI: CHEBI:86477 IUPAC Name: disodium;sulfite SMILES: [O-]S(=O)[O-].[Na+].[Na+]
Sodium Sulfite Anhydrous, Analytical Grade, Penta Chemicals
CAS: 7757-83-7 Molecular Weight (g/mol): 126.04
Sodium Sulfite, puriss. p.a., Solstice
CAS: 7757-83-7 Molecular Formula: Na2O3S Molecular Weight (g/mol): 126.037 MDL Number: MFCD00003503 InChI Key: GEHJYWRUCIMESM-UHFFFAOYSA-L Synonym: sodium sulfite,disodium sulfite,sodium sulphite,sodium sulfite anhydrous,sulfurous acid, disodium salt,sulftech,natrium sulfurosum,sodiumsulfite,natriumsulfid,natriumsulfit PubChem CID: 24437 ChEBI: CHEBI:86477 IUPAC Name: disodium;sulfite SMILES: [O-]S(=O)[O-].[Na+].[Na+]
- 1
- 2