CAS RN 7758-01-2
CAS RN 7758-01-2
Potassium bromate, Acculute Standard Volumetric Solution, Final Concentration 0.1N, Thermo Scientific Chemicals
Molecular Formula: BrKO3 Molecular Weight (g/mol): 167.00 MDL Number: MFCD00011359 InChI Key: OCATYIAKPYKMPG-UHFFFAOYSA-M Synonym: potassium bromate,bromic acid, potassium salt,bromate, potassium,potassium bromate dot,unii-04mb35w6za,ccris 529,eec no. e924,bromic acid, potassium salt 1:1,potassiumbromate,kaliumbromat PubChem CID: 23673461 ChEBI: CHEBI:38211 IUPAC Name: potassium;bromate SMILES: [O-]Br(=O)=O.[K+]
Potassium bromate concentrate, For 1L standard solution, 1/60 M KBrO3 (0.1N), Honeywell Fluka™
Molecular Formula: BrKO3 Molecular Weight (g/mol): 167.00 MDL Number: MFCD00011359 InChI Key: OCATYIAKPYKMPG-UHFFFAOYSA-M Synonym: potassium bromate,bromic acid, potassium salt,bromate, potassium,potassium bromate dot,unii-04mb35w6za,ccris 529,eec no. e924,bromic acid, potassium salt 1:1,potassiumbromate,kaliumbromat PubChem CID: 23673461 ChEBI: CHEBI:38211 IUPAC Name: potassium;bromate SMILES: [O-]Br(=O)=O.[K+]