accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2NOO

CAS RN 776-34-1

IUPAC Name: 4-nitronaphthalen-1-amine
Synonyms: 4-nitronaphthalen-1-amine
Molecular Weight (g/mol): 188.19
Molecular Formula: C10H8N2O2
InChi Key: BVPJPRYNQHAOPQ-UHFFFAOYSA-N
SMILES: NC1=C2C=CC=CC2=C(C=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)