CAS RN 7761-88-8
Silver Nitrate Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Specification Of pH.Eur., Bp, Usp, Fisher Chemical™
CAS: 7761-88-8 Molecular Formula: AgNO3 Molecular Weight (g/mol): 169.87 MDL Number: MFCD00003414 InChI Key: SQGYOTSLMSWVJD-UHFFFAOYSA-N Synonym: silver nitrate,silvernitrate,lunar caustic,silbernitrat,argenti nitras,nitrate d'argent,nitric acid silver i salt,silver mononitrate,silver i nitrate,argerol PubChem CID: 24470 ChEBI: CHEBI:32130 IUPAC Name: silver(1+) nitrate SMILES: [Ag+].[O-][N+]([O-])=O
Silver Nitrate Solution 0.05M (0.05N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
CAS: 7761-88-8 Molecular Formula: AgNO3 Molecular Weight (g/mol): 169.87 MDL Number: MFCD00003414 InChI Key: SQGYOTSLMSWVJD-UHFFFAOYSA-N Synonym: silver nitrate,silvernitrate,lunar caustic,silbernitrat,argenti nitras,nitrate d'argent,nitric acid silver i salt,silver mononitrate,silver i nitrate,argerol PubChem CID: 24470 ChEBI: CHEBI:32130 SMILES: [Ag+].[O-][N+]([O-])=O
Silver Nitrate, Certified AR for Analysis, Fisher Chemical™
CAS: 7761-88-8 Molecular Formula: AgNO3 Molecular Weight (g/mol): 169.87 MDL Number: MFCD00003414 InChI Key: SQGYOTSLMSWVJD-UHFFFAOYSA-N Synonym: silver nitrate,silvernitrate,lunar caustic,silbernitrat,argenti nitras,nitrate d'argent,nitric acid silver i salt,silver mononitrate,silver i nitrate,argerol PubChem CID: 24470 ChEBI: CHEBI:32130 SMILES: [Ag+].[O-][N+]([O-])=O
Silver Nitrate Solution 0.02M (0.02N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
CAS: 7761-88-8 Molecular Formula: AgNO3 Molecular Weight (g/mol): 169.87 MDL Number: MFCD00003414 InChI Key: SQGYOTSLMSWVJD-UHFFFAOYSA-N Synonym: silver nitrate,silvernitrate,lunar caustic,silbernitrat,argenti nitras,nitrate d'argent,nitric acid silver i salt,silver mononitrate,silver i nitrate,argerol PubChem CID: 24470 ChEBI: CHEBI:32130 SMILES: [Ag+].[O-][N+]([O-])=O
Silver nitrate, 0.1 N standard solution
CAS: 7761-88-8 | AgNO3 | 169.87 g/mol
Silver nitrate, 99.85%, for analysis
CAS: 7761-88-8 Molecular Formula: AgNO3 Molecular Weight (g/mol): 169.87 MDL Number: MFCD00003414 InChI Key: SQGYOTSLMSWVJD-UHFFFAOYSA-N Synonym: silver nitrate,silvernitrate,lunar caustic,silbernitrat,argenti nitras,nitrate d'argent,nitric acid silver i salt,silver mononitrate,silver i nitrate,argerol PubChem CID: 24470 ChEBI: CHEBI:32130 SMILES: [Ag+].[O-][N+]([O-])=O
Silver Nitrate, 99.8+%, Extra Pure, SLR, Fisher Chemical™
CAS: 7761-88-8 Molecular Formula: AgNO3 Molecular Weight (g/mol): 169.87 MDL Number: MFCD00003414 InChI Key: SQGYOTSLMSWVJD-UHFFFAOYSA-N Synonym: silver nitrate,silvernitrate,lunar caustic,silbernitrat,argenti nitras,nitrate d'argent,nitric acid silver i salt,silver mononitrate,silver i nitrate,argerol PubChem CID: 24470 ChEBI: CHEBI:32130 SMILES: [Ag+].[O-][N+]([O-])=O
Silver nitrate, 99+%, ACS reagent
CAS: 7761-88-8 Molecular Formula: AgNO3 Molecular Weight (g/mol): 169.87 MDL Number: MFCD00003414 InChI Key: SQGYOTSLMSWVJD-UHFFFAOYSA-N Synonym: silver nitrate,silvernitrate,lunar caustic,silbernitrat,argenti nitras,nitrate d'argent,nitric acid silver i salt,silver mononitrate,silver i nitrate,argerol PubChem CID: 24470 ChEBI: CHEBI:32130 IUPAC Name: silver(1+) nitrate SMILES: [Ag+].[O-][N+]([O-])=O
Silver Nitrate (Crystalline/Certified ACS), Fisher Chemical™
CAS: 7761-88-8 Molecular Formula: AgNO3 Molecular Weight (g/mol): 169.87 MDL Number: MFCD00003414 InChI Key: SQGYOTSLMSWVJD-UHFFFAOYSA-N Synonym: silver nitrate,silvernitrate,lunar caustic,silbernitrat,argenti nitras,nitrate d'argent,nitric acid silver i salt,silver mononitrate,silver i nitrate,argerol PubChem CID: 24470 ChEBI: CHEBI:32130 IUPAC Name: silver(1+) nitrate SMILES: [Ag+].[O-][N+]([O-])=O
Silver nitrate, ACS, 99.9+% (metals basis)
CAS: 7761-88-8 Molecular Formula: AgNO3 Molecular Weight (g/mol): 169.87 MDL Number: MFCD00003414 InChI Key: SQGYOTSLMSWVJD-UHFFFAOYSA-N Synonym: silver nitrate,silvernitrate,lunar caustic,silbernitrat,argenti nitras,nitrate d'argent,nitric acid silver i salt,silver mononitrate,silver i nitrate,argerol PubChem CID: 24470 ChEBI: CHEBI:32130 IUPAC Name: silver(1+) nitrate SMILES: [Ag+].[O-][N+]([O-])=O
Silver nitrate, Premion™, 99.995% (metals basis), Ag 63% min
CAS: 7761-88-8 Molecular Formula: AgNO3 Molecular Weight (g/mol): 169.87 MDL Number: MFCD00003414 InChI Key: SQGYOTSLMSWVJD-UHFFFAOYSA-N Synonym: silver nitrate,silvernitrate,lunar caustic,silbernitrat,argenti nitras,nitrate d'argent,nitric acid silver i salt,silver mononitrate,silver i nitrate,argerol PubChem CID: 24470 ChEBI: CHEBI:32130 IUPAC Name: silver(1+) nitrate SMILES: [Ag+].[O-][N+]([O-])=O
Silver nitrate, Premion™, 99.9995% (metals basis)
CAS: 7761-88-8 Molecular Formula: AgNO3 Molecular Weight (g/mol): 169.87 MDL Number: MFCD00003414 InChI Key: SQGYOTSLMSWVJD-UHFFFAOYSA-N Synonym: silver nitrate,silvernitrate,lunar caustic,silbernitrat,argenti nitras,nitrate d'argent,nitric acid silver i salt,silver mononitrate,silver i nitrate,argerol PubChem CID: 24470 ChEBI: CHEBI:32130 IUPAC Name: silver(1+) nitrate SMILES: [Ag+].[O-][N+]([O-])=O
Silver standard solution, for AAS, 1 mg/ml Ag in 0.5N HNO3
CAS: 7761-88-8 | AgNO3 | 169.87 g/mol