accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClClSn

CAS RN 7772-99-8

IUPAC Name: λ²-tin(2+) dichloride
Synonyms: stannous chloride
Molecular Weight (g/mol): 189.61
Molecular Formula: Cl2Sn
InChi Key: AXZWODMDQAVCJE-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Sn++]
Molecular Weight (g/mol) 
  • (1)
  • (23)
Quantity 
  • (1)
  • (2)
  • (1)
  • (6)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
Physical Form 
  • (6)
  • (1)
  • (4)
  • (4)
  • (1)
Percent Purity 
  • (1)
  • (10)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (4)
  • (4)
  • (10)
  • (1)
  • (1)
  • (23)
  • (1)
  • (2)
  • (1)
  • (6)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
  • (6)
  • (1)
  • (4)
  • (4)
  • (1)
  • (1)
  • (10)
  • (3)
  • (1)