accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH4NH2SOOO

CAS RN 7773-06-0

IUPAC Name: ammonium sulfamate
Synonyms: ammonium sulfamate
Molecular Weight (g/mol): 114.12
Molecular Formula: H6N2O3S
InChi Key: GEHMBYLTCISYNY-UHFFFAOYSA-N
SMILES: [NH4+].NS([O-])(=O)=O
Quantity 
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (4)
  • (7)
  • (1)
Grade 
  • (9)
  • (1)
Physical Form 
  • (4)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (4)
  • (4)
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
  • (7)
  • (1)
  • (9)
  • (1)
  • (4)
  • (4)