accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NaNaOSOSOO

CAS RN 7775-14-6

IUPAC Name: disodium sulfinatosulfinate
Synonyms: sodium dithionite sodium hydrosulfite
Molecular Weight (g/mol): 174.1
Molecular Formula: Na2O4S2
InChi Key: JVBXVOWTABLYPX-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]S(=O)S([O-])=O
Molecular Weight (g/mol) 
  • (12)
  • (2)
  • (3)
Quantity 
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
Grade 
  • (9)
Physical Form 
  • (4)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (4)
  • (3)
  • (3)
  • (12)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (9)
  • (4)
  • (3)
  • (2)
  • (4)