accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7775-41-9

FAg

CAS RN 7775-41-9

IUPAC Name: silver(1+) fluoride
Synonyms: AgF silver monofluoride argentous fluoride silver(I) fluoride
Molecular Weight (g/mol): 126.87
Molecular Formula: AgF
InChi Key: REYHXKZHIMGNSE-UHFFFAOYSA-M
SMILES: [F-].[Ag+]
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (4)
Physical Form 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (5)
  • (1)
  • (1)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)