accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7778-18-9

IUPAC Name: calcium sulfate
Synonyms: calcium sulfate CaSO4 kalziumsulfat calcium sulfate, anhydrous anhydrous gypsum
Molecular Weight (g/mol): 136.13
Molecular Formula: CaO4S
InChi Key: OSGAYBCDTDRGGQ-UHFFFAOYSA-L
SMILES: [Ca++].[O-]S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (58)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (7)
  • (2)
  • (1)
  • (1)
  • (8)
  • (5)
  • (1)
  • (7)
  • (1)
  • (1)
  • (6)
  • (1)
  • (2)
  • (1)
  • (12)
Physical Form 
  • (4)
  • (6)
  • (4)
Grade 
  • (6)
  • (3)
  • (4)
Material 
  • (10)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (12)
  • (1)
  • (28)
  • (13)
  • (3)
  • (5)
  • (58)
  • (3)
  • (1)
  • (2)
  • (2)
  • (7)
  • (2)
  • (1)
  • (1)
  • (8)
  • (5)
  • (1)
  • (7)
  • (1)
  • (1)
  • (6)
  • (1)
  • (2)
  • (1)
  • (12)
  • (4)
  • (6)
  • (4)
  • (6)
  • (3)
  • (4)
  • (10)
  • (3)
  • (6)
  • (3)