accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

KOClOOO

CAS RN 7778-74-7

IUPAC Name: potassium perchlorate
Synonyms: potassium perchlorate
Molecular Weight (g/mol): 138.54
Molecular Formula: ClKO4
InChi Key: YLMGFJXSLBMXHK-UHFFFAOYSA-M
SMILES: [K+].[O-][Cl](=O)(=O)=O
Quantity 
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (6)
Percent Purity 
  • (2)
  • (4)
  • (3)
  • (2)
Grade 
  • (2)
  • (4)
  • (4)
Physical Form 
  • (2)
  • (4)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (6)
  • (5)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (6)
  • (2)
  • (4)
  • (3)
  • (2)
  • (2)
  • (4)
  • (4)
  • (2)
  • (4)
  • (2)
  • (1)