CAS RN 7778-77-0
Potassium Dihydrogen Orthophosphate, Certified AR for Analysis, Fisher Chemical™
CAS: 7778-77-0 Molecular Formula: H2KO4P Molecular Weight (g/mol): 136.08 MDL Number: MFCD00011401 MFCD00147253 InChI Key: GNSKLFRGEWLPPA-UHFFFAOYSA-M Synonym: potassium dihydrogen phosphate,monopotassium phosphate,potassium phosphate monobasic,potassium phosphate, monobasic,potassium acid phosphate,phosphoric acid, monopotassium salt,monobasic potassium phosphate,monopotassium monophosphate,monopotassium dihydrogen phosphate,monopotassium orthophosphate PubChem CID: 516951 ChEBI: CHEBI:63036 SMILES: [K+].OP(O)([O-])=O
Potassium phosphate, monobasic, 99+%, ACS reagent
CAS: 7778-77-0 Molecular Formula: H2KO4P Molecular Weight (g/mol): 136.08 MDL Number: MFCD00011401 MFCD00147253 InChI Key: GNSKLFRGEWLPPA-UHFFFAOYSA-M Synonym: potassium dihydrogen phosphate,monopotassium phosphate,potassium phosphate monobasic,potassium phosphate, monobasic,potassium acid phosphate,phosphoric acid, monopotassium salt,monobasic potassium phosphate,monopotassium monophosphate,monopotassium dihydrogen phosphate,monopotassium orthophosphate PubChem CID: 516951 ChEBI: CHEBI:63036 SMILES: [K+].OP(O)([O-])=O
Buffer Solution pH6.00 (Phosphate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical™
Buffer Solution pH6.00 (Phosphate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical, Quantity: 1L, CAS: 7778-77-0, Colorless, Packaging: HDPE plastic bottle, pH: 6, MDL Number: 81857, Physical Form: Liquid
Potassium dihydrogen phosphate, 98+%
CAS: 7778-77-0 Molecular Formula: H2KO4P Molecular Weight (g/mol): 136.08 MDL Number: MFCD00011401 MFCD00147253 InChI Key: GNSKLFRGEWLPPA-UHFFFAOYSA-M Synonym: potassium dihydrogen phosphate,monopotassium phosphate,potassium phosphate monobasic,potassium phosphate, monobasic,potassium acid phosphate,phosphoric acid, monopotassium salt,monobasic potassium phosphate,monopotassium monophosphate,monopotassium dihydrogen phosphate,monopotassium orthophosphate PubChem CID: 516951 ChEBI: CHEBI:63036 IUPAC Name: potassium dihydrogen phosphate SMILES: [K+].OP(O)([O-])=O
Potassium Dihydrogen Orthophosphate, for HPLC, Fisher Chemical™
CAS: 7778-77-0 Molecular Formula: H2KO4P Molecular Weight (g/mol): 136.08 MDL Number: MFCD00011401 MFCD00147253 InChI Key: GNSKLFRGEWLPPA-UHFFFAOYSA-M Synonym: potassium dihydrogen phosphate,monopotassium phosphate,potassium phosphate monobasic,potassium phosphate, monobasic,potassium acid phosphate,phosphoric acid, monopotassium salt,monobasic potassium phosphate,monopotassium monophosphate,monopotassium dihydrogen phosphate,monopotassium orthophosphate PubChem CID: 516951 ChEBI: CHEBI:63036 SMILES: [K+].OP(O)([O-])=O
Potassium phosphate, monobasic, for HPLC
CAS: 7778-77-0 Molecular Formula: H2KO4P Molecular Weight (g/mol): 136.08 MDL Number: MFCD00011401 MFCD00147253 InChI Key: GNSKLFRGEWLPPA-UHFFFAOYSA-M IUPAC Name: potassium dihydrogen phosphate SMILES: [K+].OP(O)([O-])=O
Potassium dihydrogen phosphate, ACS, 99.0% min
CAS: 7778-77-0 Molecular Formula: H2KO4P Molecular Weight (g/mol): 136.08 MDL Number: MFCD00011401 MFCD00147253 InChI Key: GNSKLFRGEWLPPA-UHFFFAOYSA-M Synonym: potassium dihydrogen phosphate,monopotassium phosphate,potassium phosphate monobasic,potassium phosphate, monobasic,potassium acid phosphate,phosphoric acid, monopotassium salt,monobasic potassium phosphate,monopotassium monophosphate,monopotassium dihydrogen phosphate,monopotassium orthophosphate PubChem CID: 516951 ChEBI: CHEBI:63036 SMILES: [K+].OP(O)([O-])=O
Potassium Dihydrogen Orthophosphate, Extra Pure, SLR, Fisher Chemical™
CAS: 7778-77-0 Molecular Formula: H2KO4P Molecular Weight (g/mol): 136.08 MDL Number: MFCD00011401 MFCD00147253 InChI Key: GNSKLFRGEWLPPA-UHFFFAOYSA-M Synonym: potassium dihydrogen phosphate,monopotassium phosphate,potassium phosphate monobasic,potassium phosphate, monobasic,potassium acid phosphate,phosphoric acid, monopotassium salt,monobasic potassium phosphate,monopotassium monophosphate,monopotassium dihydrogen phosphate,monopotassium orthophosphate PubChem CID: 516951 ChEBI: CHEBI:63036 SMILES: [K+].OP(O)([O-])=O
Thermo Scientific Chemicals Potassium phosphate, monobasic, 99+%, for biochemistry
CAS: 7778-77-0 Molecular Formula: H2KO4P Molecular Weight (g/mol): 136.08 MDL Number: MFCD00011401 MFCD00147253 InChI Key: GNSKLFRGEWLPPA-UHFFFAOYSA-M Synonym: potassium dihydrogen phosphate,monopotassium phosphate,potassium phosphate monobasic,potassium phosphate, monobasic,potassium acid phosphate,phosphoric acid, monopotassium salt,monobasic potassium phosphate,monopotassium monophosphate,monopotassium dihydrogen phosphate,monopotassium orthophosphate PubChem CID: 516951 ChEBI: CHEBI:63036 SMILES: [K+].OP(O)([O-])=O
Potassium Phosphate Monobasic (White Crystals), Fisher BioReagents
CAS: 7778-77-0 Molecular Formula: H2KO4P Molecular Weight (g/mol): 136.08 MDL Number: MFCD00011401 MFCD00147253 InChI Key: GNSKLFRGEWLPPA-UHFFFAOYSA-M Synonym: potassium dihydrogen phosphate,monopotassium phosphate,potassium phosphate monobasic,potassium phosphate, monobasic,potassium acid phosphate,phosphoric acid, monopotassium salt,monobasic potassium phosphate,monopotassium monophosphate,monopotassium dihydrogen phosphate,monopotassium orthophosphate PubChem CID: 516951 ChEBI: CHEBI:63036 SMILES: [K+].OP(O)([O-])=O
Potassium phosphate, monobasic, 99+%, pure
CAS: 7778-77-0 Molecular Formula: H2KO4P Molecular Weight (g/mol): 136.08 MDL Number: MFCD00011401 MFCD00147253 InChI Key: GNSKLFRGEWLPPA-UHFFFAOYSA-M Synonym: potassium dihydrogen phosphate,monopotassium phosphate,potassium phosphate monobasic,potassium phosphate, monobasic,potassium acid phosphate,phosphoric acid, monopotassium salt,monobasic potassium phosphate,monopotassium monophosphate,monopotassium dihydrogen phosphate,monopotassium orthophosphate PubChem CID: 516951 ChEBI: CHEBI:63036 SMILES: [K+].OP(O)([O-])=O
Buffer Colour Coded Solution pH7.00 (Phosphate) Yellow, NIST Standard Solution ready to use for pH measurement, Fisher Chemical™
CAS: 7778-77-0 Molecular Formula: H2KO4P Molecular Weight (g/mol): 136.08 InChI Key: GNSKLFRGEWLPPA-UHFFFAOYSA-M IUPAC Name: potassium dihydrogen phosphate SMILES: [K+].OP(O)([O-])=O
Potassium phosphate, monobasic, 99+%, extra pure
CAS: 7778-77-0 Molecular Formula: H2KO4P Molecular Weight (g/mol): 136.08 MDL Number: MFCD00011401 MFCD00147253 InChI Key: GNSKLFRGEWLPPA-UHFFFAOYSA-M Synonym: potassium dihydrogen phosphate,monopotassium phosphate,potassium phosphate monobasic,potassium phosphate, monobasic,potassium acid phosphate,phosphoric acid, monopotassium salt,monobasic potassium phosphate,monopotassium monophosphate,monopotassium dihydrogen phosphate,monopotassium orthophosphate PubChem CID: 516951 ChEBI: CHEBI:63036 IUPAC Name: potassium dihydrogen phosphate SMILES: [K+].OP(O)([O-])=O
Potassium phosphate monobasic, puriss. p.a., ACS Reagent, Solstice
CAS: 7778-77-0 Molecular Formula: H2KO4P Molecular Weight (g/mol): 136.08 MDL Number: MFCD00011401 MFCD00147253 InChI Key: GNSKLFRGEWLPPA-UHFFFAOYSA-M Synonym: potassium dihydrogen phosphate,monopotassium phosphate,potassium phosphate monobasic,potassium phosphate, monobasic,potassium acid phosphate,phosphoric acid, monopotassium salt,monobasic potassium phosphate,monopotassium monophosphate,monopotassium dihydrogen phosphate,monopotassium orthophosphate PubChem CID: 516951 ChEBI: CHEBI:63036 IUPAC Name: potassium;dihydrogen phosphate SMILES: [K+].OP(O)([O-])=O
Potassium phosphate monobasic, meets analytical spec. of NF, Ph. Eur., E340, Anhydrous, 98-100.5% (calc. to the dried substance), Solstice
CAS: 7778-77-0 Molecular Formula: H2KO4P Molecular Weight (g/mol): 136.08 MDL Number: MFCD00011401 MFCD00147253 InChI Key: GNSKLFRGEWLPPA-UHFFFAOYSA-M Synonym: potassium dihydrogen phosphate,monopotassium phosphate,potassium phosphate monobasic,potassium phosphate, monobasic,potassium acid phosphate,phosphoric acid, monopotassium salt,monobasic potassium phosphate,monopotassium monophosphate,monopotassium dihydrogen phosphate,monopotassium orthophosphate PubChem CID: 516951 ChEBI: CHEBI:63036 SMILES: [K+].OP(O)([O-])=O