accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3OO(E)

CAS RN 7780-06-5

IUPAC Name: propan-2-yl (2E)-3-phenylprop-2-enoate
Synonyms: isopropyl cinnamate
Molecular Weight (g/mol): 190.24
Molecular Formula: C12H14O2
InChi Key: RGACABDFLVLVCT-CMDGGOBGSA-N
SMILES: CC(C)OC(=O)\C=C\C1=CC=CC=C1
Boiling Point 
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results