CAS RN 7783-20-2
Ammonium sulfate, 99+%, ACS reagent
CAS: 7783-20-2 Molecular Formula: H8N2O4S Molecular Weight (g/mol): 132.13 MDL Number: MFCD00003391 InChI Key: BFNBIHQBYMNNAN-UHFFFAOYSA-N Synonym: ammonium sulfate,diammonium sulfate,ammonium sulphate,mascagnite,sulfuric acid diammonium salt,ammonium sulfate 2:1,ammoniumsulfate,actamaster,sulfuric acid, diammonium salt,diammonium sulphate PubChem CID: 6097028 ChEBI: CHEBI:62946 SMILES: N.N.OS(O)(=O)=O
Ammonium sulfate, 99.5%, for analysis
CAS: 7783-20-2 Molecular Formula: H8N2O4S Molecular Weight (g/mol): 132.13 MDL Number: MFCD00003391 InChI Key: BFNBIHQBYMNNAN-UHFFFAOYSA-N Synonym: ammonium sulfate,diammonium sulfate,ammonium sulphate,mascagnite,sulfuric acid diammonium salt,ammonium sulfate 2:1,ammoniumsulfate,actamaster,sulfuric acid, diammonium salt,diammonium sulphate PubChem CID: 6097028 ChEBI: CHEBI:62946 SMILES: N.N.OS(O)(=O)=O
Ammonium sulfate, ultrapure, 99+%
CAS: 7783-20-2 Molecular Formula: H8N2O4S Molecular Weight (g/mol): 132.13 MDL Number: MFCD00003391 InChI Key: BFNBIHQBYMNNAN-UHFFFAOYSA-N Synonym: ammonium sulfate,diammonium sulfate,ammonium sulphate,mascagnite,sulfuric acid diammonium salt,ammonium sulfate 2:1,ammoniumsulfate,actamaster,sulfuric acid, diammonium salt,diammonium sulphate PubChem CID: 6097028 ChEBI: CHEBI:62946 IUPAC Name: sulfuric acid diamine SMILES: N.N.OS(O)(=O)=O
Ammonium sulfate, 99%, pure
CAS: 7783-20-2 Molecular Formula: H8N2O4S Molecular Weight (g/mol): 132.13 MDL Number: MFCD00003391 InChI Key: BFNBIHQBYMNNAN-UHFFFAOYSA-N Synonym: ammonium sulfate,diammonium sulfate,ammonium sulphate,mascagnite,sulfuric acid diammonium salt,ammonium sulfate 2:1,ammoniumsulfate,actamaster,sulfuric acid, diammonium salt,diammonium sulphate PubChem CID: 6097028 ChEBI: CHEBI:62946 SMILES: N.N.OS(O)(=O)=O
Ammonium sulfate, ACS, 99.0% min
CAS: 7783-20-2 Molecular Formula: H8N2O4S Molecular Weight (g/mol): 132.13 MDL Number: MFCD00003391 InChI Key: BFNBIHQBYMNNAN-UHFFFAOYSA-N Synonym: ammonium sulfate,diammonium sulfate,ammonium sulphate,mascagnite,sulfuric acid diammonium salt,ammonium sulfate 2:1,ammoniumsulfate,actamaster,sulfuric acid, diammonium salt,diammonium sulphate PubChem CID: 6097028 ChEBI: CHEBI:62946 SMILES: N.N.OS(O)(=O)=O
Ammonium sulfate, 98+%
CAS: 7783-20-2 Molecular Formula: H8N2O4S Molecular Weight (g/mol): 132.13 MDL Number: MFCD00003391 InChI Key: BFNBIHQBYMNNAN-UHFFFAOYSA-N Synonym: ammonium sulfate,diammonium sulfate,ammonium sulphate,mascagnite,sulfuric acid diammonium salt,ammonium sulfate 2:1,ammoniumsulfate,actamaster,sulfuric acid, diammonium salt,diammonium sulphate PubChem CID: 6097028 ChEBI: CHEBI:62946 IUPAC Name: diazanium;sulfate SMILES: N.N.OS(O)(=O)=O
Ammonium Sulfate, Certified AR for Analysis, Fisher Chemical™
CAS: 7783-20-2 Molecular Formula: H8N2O4S Molecular Weight (g/mol): 132.13 MDL Number: MFCD00003391 InChI Key: BFNBIHQBYMNNAN-UHFFFAOYSA-N IUPAC Name: sulfuric acid diamine SMILES: N.N.OS(O)(=O)=O
Ammonium Sulfate, 99%, extra pure
CAS: 7783-20-2 Molecular Formula: H8N2O4S Molecular Weight (g/mol): 132.13 MDL Number: MFCD00003391 InChI Key: BFNBIHQBYMNNAN-UHFFFAOYSA-N Synonym: ammonium sulfate,diammonium sulfate,ammonium sulphate,mascagnite,sulfuric acid diammonium salt,ammonium sulfate 2:1,ammoniumsulfate,actamaster,sulfuric acid, diammonium salt,diammonium sulphate PubChem CID: 6097028 ChEBI: CHEBI:62946 IUPAC Name: sulfuric acid diamine SMILES: N.N.OS(O)(=O)=O
Ammonium sulfate, 99.95% (metals basis)
CAS: 7783-20-2 Molecular Formula: H8N2O4S Molecular Weight (g/mol): 132.13 MDL Number: MFCD00003391 InChI Key: BFNBIHQBYMNNAN-UHFFFAOYSA-N Synonym: ammonium sulfate,diammonium sulfate,ammonium sulphate,mascagnite,sulfuric acid diammonium salt,ammonium sulfate 2:1,ammoniumsulfate,actamaster,sulfuric acid, diammonium salt,diammonium sulphate PubChem CID: 6097028 ChEBI: CHEBI:62946 IUPAC Name: sulfuric acid diamine SMILES: N.N.OS(O)(=O)=O
Ammonium sulfate, 99%, for biochemistry
CAS: 7783-20-2 Molecular Formula: H8N2O4S Molecular Weight (g/mol): 132.13 MDL Number: MFCD00003391 InChI Key: BFNBIHQBYMNNAN-UHFFFAOYSA-N Synonym: ammonium sulfate,diammonium sulfate,ammonium sulphate,mascagnite,sulfuric acid diammonium salt,ammonium sulfate 2:1,ammoniumsulfate,actamaster,sulfuric acid, diammonium salt,diammonium sulphate PubChem CID: 6097028 ChEBI: CHEBI:62946 IUPAC Name: diazanium;sulfate SMILES: N.N.OS(O)(=O)=O
Ammonium sulfate, 99.9995%, (trace metal basis)
CAS: 7783-20-2 Molecular Formula: H8N2O4S Molecular Weight (g/mol): 132.13 MDL Number: MFCD00003391 InChI Key: BFNBIHQBYMNNAN-UHFFFAOYSA-N Synonym: ammonium sulfate,diammonium sulfate,ammonium sulphate,mascagnite,sulfuric acid diammonium salt,ammonium sulfate 2:1,ammoniumsulfate,actamaster,sulfuric acid, diammonium salt,diammonium sulphate PubChem CID: 6097028 ChEBI: CHEBI:62946 IUPAC Name: diazanium;sulfate SMILES: N.N.OS(O)(=O)=O
Ammonium sulfate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99%, Solstice
CAS: 7783-20-2 Molecular Formula: H8N2O4S Molecular Weight (g/mol): 132.13 MDL Number: MFCD00003391 InChI Key: BFNBIHQBYMNNAN-UHFFFAOYSA-N Synonym: ammonium sulfate,diammonium sulfate,ammonium sulphate,mascagnite,sulfuric acid diammonium salt,ammonium sulfate 2:1,ammoniumsulfate,actamaster,sulfuric acid, diammonium salt,diammonium sulphate PubChem CID: 6097028 ChEBI: CHEBI:62946 IUPAC Name: diazanium;sulfate SMILES: N.N.OS(O)(=O)=O
Ammonium Sulfate, Fisher BioReagents
CAS: 7783-20-2 Molecular Formula: H8N2O4S Molecular Weight (g/mol): 132.13 MDL Number: MFCD00003391 InChI Key: BFNBIHQBYMNNAN-UHFFFAOYSA-N Synonym: ammonium sulfate,diammonium sulfate,ammonium sulphate,mascagnite,sulfuric acid diammonium salt,ammonium sulfate 2:1,ammoniumsulfate,actamaster,sulfuric acid, diammonium salt,diammonium sulphate PubChem CID: 6097028 ChEBI: CHEBI:62946 IUPAC Name: sulfuric acid diamine SMILES: N.N.OS(O)(=O)=O
Ammonium sulfate, Puratronic™, 99.999% (metals basis)
CAS: 7783-20-2 Molecular Formula: H8N2O4S Molecular Weight (g/mol): 132.13 MDL Number: MFCD00003391 InChI Key: BFNBIHQBYMNNAN-UHFFFAOYSA-N Synonym: ammonium sulfate,diammonium sulfate,ammonium sulphate,mascagnite,sulfuric acid diammonium salt,ammonium sulfate 2:1,ammoniumsulfate,actamaster,sulfuric acid, diammonium salt,diammonium sulphate PubChem CID: 6097028 ChEBI: CHEBI:62946 SMILES: N.N.OS(O)(=O)=O