accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONOOONOOHgH2O

CAS RN 7783-34-8

IUPAC Name: mercury(2+) hydrate dinitrate
Synonyms: mercury(2+) hydrate dinitrate
Molecular Weight (g/mol): 342.61
Molecular Formula: H2HgN2O7
InChi Key: KVICROHOONHSRH-UHFFFAOYSA-N
SMILES: O.[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)
  • (1)
Grade 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)