accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFFFe

CAS RN 7783-50-8

IUPAC Name: iron(3+) trifluoride
Synonyms: iron(III) fluoride
Molecular Weight (g/mol): 112.84
Molecular Formula: F3Fe
InChi Key: SHXXPRJOPFJRHA-UHFFFAOYSA-K
SMILES: [F-].[F-].[F-].[Fe+3]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results