accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

SbFFF

CAS RN 7783-56-4

IUPAC Name: antimony(3+) trifluoride
Synonyms: antimony trifluoride
Molecular Weight (g/mol): 178.76
Molecular Formula: F3Sb
InChi Key: GUNJVIDCYZYFGV-UHFFFAOYSA-K
SMILES: [F-].[F-].[F-].[Sb+3]
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (2)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)