accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH4H2OH2OH2OH2OH2OH2OH2OH2OH2OH2OH2OH2OFeOSOOOOSOOO

CAS RN 7783-83-7

IUPAC Name: iron(3+) ammonium dodecahydrate disulfate
Synonyms: iron(3+) ammonium dodecahydrate disulfate
Molecular Weight (g/mol): 482.18
Molecular Formula: FeH28NO20S2
InChi Key: LCPUDZUWZDSKMX-UHFFFAOYSA-K
SMILES: [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (13)
  • (5)
  • (8)
  • (3)
Quantity 
  • (2)
  • (2)
  • (5)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (11)
Grade 
  • (3)
  • (7)
  • (1)
  • (2)
  • (5)
  • (1)
  • (1)
Physical Form 
  • (4)
  • (2)
  • (3)
  • (1)
  • (1)
  • (4)
  • (4)
Percent Purity 
  • (2)
  • (5)
  • (2)
  • (6)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (3)
  • (3)
  • (2)
  • (1)
  • (7)
  • (4)
  • (3)
  • (1)
  • (4)
  • (13)
  • (5)
  • (8)
  • (3)
  • (2)
  • (2)
  • (5)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (11)
  • (3)
  • (7)
  • (1)
  • (2)
  • (5)
  • (1)
  • (1)
  • (4)
  • (2)
  • (3)
  • (1)
  • (1)
  • (4)
  • (4)
  • (2)
  • (5)
  • (2)
  • (6)
  • (1)
  • (1)
  • (1)