accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

AgCl

CAS RN 7783-90-6

IUPAC Name: silver(1+) chloride
Synonyms: silver chloride silver monochloride AgCl silver chloride deposited on titanium dioxide
Molecular Weight (g/mol): 143.32
Molecular Formula: AgCl
InChi Key: HKZLPVFGJNLROG-UHFFFAOYSA-M
SMILES: [Cl-].[Ag+]
Quantity 
  • (1)
  • (1)
  • (2)
  • (6)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (4)
  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
Physical Form 
  • (2)
  • (5)
  • (3)
  • (4)
  • (2)
Grade 
  • (4)
  • (3)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (4)
  • (13)
  • (1)
  • (1)
  • (2)
  • (6)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (2)
  • (5)
  • (3)
  • (4)
  • (2)
  • (4)
  • (3)
  • (3)
  • (2)
  • (2)