accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7784-03-4

IIIIAgAgHg

CAS RN 7784-03-4

IUPAC Name: mercury(2+) disilver(1+) tetraiodide
Synonyms: mercury(2+) disilver(1+) tetraiodide
Molecular Weight (g/mol): 923.94
Molecular Formula: Ag2HgI4
InChi Key: YNTJRFXPYRRRKS-UHFFFAOYSA-J
SMILES: [Ag+].[Ag+].[I-].[I-].[I-].[I-].[Hg++]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results