accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7784-09-0

AgAgAgOPOOO

CAS RN 7784-09-0

IUPAC Name: trisilver(1+) phosphate
Synonyms: silver phosphate
Molecular Weight (g/mol): 418.57
Molecular Formula: Ag3O4P
InChi Key: FJOLTQXXWSRAIX-UHFFFAOYSA-K
SMILES: [Ag+].[Ag+].[Ag+].[O-]P([O-])([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (1)