accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2OH2OH2OH2OH2OH2OAlClClCl

CAS RN 7784-13-6

IUPAC Name: aluminium(3+) hexahydrate trichloride
Synonyms: aluminium(3+) hexahydrate trichloride
Molecular Weight (g/mol): 241.42
Molecular Formula: AlCl3H12O6
InChi Key: JGDITNMASUZKPW-UHFFFAOYSA-K
SMILES: O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]
Molecular Weight (g/mol) 
  • (22)
  • (3)
Quantity 
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (5)
  • (1)
Grade 
  • (4)
  • (4)
  • (3)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (9)
  • (4)
Physical Form 
  • (4)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (4)
  • (1)
  • (4)
  • (11)
  • (3)
  • (5)
  • (22)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (5)
  • (1)
  • (4)
  • (4)
  • (3)
  • (2)
  • (3)
  • (3)
  • (9)
  • (4)
  • (4)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)