accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH4MgOPOOO

CAS RN 7785-21-9

IUPAC Name: phosphoric acid amine hexahydrate magnesium
Synonyms: phosphoric acid amine hexahydrate magnesium
Molecular Weight (g/mol): 247.42
Molecular Formula: H18MgNO10P
InChi Key: QMXULYNJFFJVLV-UHFFFAOYSA-N
SMILES: N.O.O.O.O.O.O.[Mg].OP(O)(O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results