accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7785-26-4

CH3(S)(S)H3CH3C

CAS RN 7785-26-4

IUPAC Name: (1S,5S)-2,6,6-trimethylbicyclo[3.1.1]hept-2-ene
Synonyms: (-)-α-pinene (1S,5S)-α-pinene L-α-pinene
Molecular Weight (g/mol): 136.24
Molecular Formula: C10H16
InChi Key: GRWFGVWFFZKLTI-IUCAKERBSA-N
SMILES: CC1=CC[C@H]2C[C@@H]1C2(C)C
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (4)