accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7786-67-6

CH3HOCH3H2C

CAS RN 7786-67-6

IUPAC Name: 5-methyl-2-(prop-1-en-2-yl)cyclohexan-1-ol
Synonyms: 5-methyl-2-(prop-1-en-2-yl)cyclohexan-1-ol
Molecular Weight (g/mol): 154.25
Molecular Formula: C10H18O
InChi Key: ZYTMANIQRDEHIO-UHFFFAOYNA-N
SMILES: CC1CCC(C(O)C1)C(C)=C
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results