accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7787-35-1

BaOMnOOO

CAS RN 7787-35-1

IUPAC Name: barium(2+) dioxomanganesebis(olate)
Synonyms: barium manganate
Molecular Weight (g/mol): 256.26
Molecular Formula: BaMnO4
InChi Key: ZZCNKSMCIZCVDR-UHFFFAOYSA-N
SMILES: [Ba++].[O-][Mn]([O-])(=O)=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)