accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BeH2OH2OH2OH2OOSOOO

CAS RN 7787-56-6

IUPAC Name: beryllium(2+) tetrahydrate sulfate
Synonyms: beryllium sulfate tetrahydrate beryllium sulfate, tetrahydrate (1:1:4)
Molecular Weight (g/mol): 177.13
Molecular Formula: BeH8O8S
InChi Key: DIMYTQPLZWDZFE-UHFFFAOYSA-L
SMILES: [Be++].O.O.O.O.[O-]S([O-])(=O)=O
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)