accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7787-58-8

BrBrBrBi

CAS RN 7787-58-8

IUPAC Name: bismuth(3+) tribromide
Synonyms: bismuth tribromide
Molecular Weight (g/mol): 448.69
Molecular Formula: BiBr3
InChi Key: TXKAQZRUJUNDHI-UHFFFAOYSA-K
SMILES: [Br-].[Br-].[Br-].[Bi+3]
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)